Anti-Ribonuclease inhibitor 1 Mouse Monoclonal Antibody [clone: 10C309]
USBIR2025-01
- Antibody type:Primary
- Antigen name:Ribonuclease inhibitor 1
- Clonality:Monoclonal
- Clone:10C309
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:7 to 461
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000-2000
Type: Primary
Antigen: Ribonuclease inhibitor 1
Clonality: monoclonal
Clone: 10C309
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human