Anti-UPF1 Rabbit Polyclonal Antibody
USBIR1495-03H
- Antibody type:Primary
- Antigen name:UPF1 regulator of nonsense transcripts homolog (yeast)
- Antigen symbol:UPF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Form:liquid
- Amino acid number:304 to 334
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:200 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Type: Primary
Antigen: UPF1 (UPF1 regulator of nonsense transcripts homolog (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human