Anti-CCL5 Mouse Monoclonal Antibody (Biotin) [clone: 5J49]
Catalog # USBIR1100-02D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) ligand 5
- Antigen symbol:CCL5
- Clonality:Monoclonal
- Clone:5J49
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,3, 0,5% BSA, 0,05% sodium azide
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0.4-1.0 ?g/ml
Type: Primary
Antigen: CCL5 (chemokine (C-C motif) ligand 5)
Clonality: monoclonal
Clone: 5J49
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human