Anti-QKI Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # USBIQ0500-01E-APC
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Qki, Kh Domain Containing, Rna Binding
- Antigen symbol:QK1
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50
Do not freeze!
Type: Primary
Antigen: QKI (QKI, KH domain containing, RNA binding)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human