Anti-PRNP Mouse Monoclonal Antibody [clone: 6D685]
USBIP8950-02C
- Antibody type:Primary
- Antigen name:Prion Protein
- Antigen symbol:PRNP
- Clonality:Monoclonal
- Clone:6D685
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Hamster,Sheep,Human,Bovine,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Format:Supernatant
- Form:liquid
- Amino acid number:97 to 102
- Storage buffer:culture medium (phenol red and nutrients still present) supplemented with FBS and 0,01% thimerosal .
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Recommended dilution: ELISA: 1:5000-1:20000; Western Blot: 1:5000-1:20000
Type: Primary
Antigen: PRNP (prion protein)
Clonality: monoclonal
Clone: 6D685
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Hamster, Human, Mouse, Sheep