Anti-Plastin-2 Mouse Monoclonal Antibody [clone: SPM414]
Catalog # USBIP4268-01D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Plastin 2
- Antigen symbol:PLS2
- Clonality:Monoclonal
- Clone:SPM414
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,2% BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: Plastin-2
Clonality: monoclonal
Clone: SPM414
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human