Anti-Phosphotyrosine Mouse Monoclonal Antibody [clone: 9F636]
USBIP4079G
- Antibody type:Primary
- Antigen name:Phosphotyrosine
- Antigen symbol:pY
- Clonality:Monoclonal
- Clone:9F636
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 1:4000; Western Blot: 1:4000; Immunochemistry: 1:4000; Immunoprecipitation: 1:4000
Type: Primary
Antigen: Phosphotyrosine
Clonality: monoclonal
Clone: 9F636
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: