Anti-ALPL Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
USBIP4071-18B-AP
- Antibody type:Primary
- Antigen name:Alkaline Phosphatase, liver/bone/kidney
- Antigen symbol:ALPL
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Form:liquid
- Amino acid number:10 to 40
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: ALPL (alkaline phosphatase, liver/bone/kidney)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse