Anti-OGT Goat Polyclonal Antibody
Catalog # USBIO5750-01B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:O-linked N-acetylglucosamine (GlcNAc) transferase
- Antigen symbol:OGT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Form:liquid
- Storage buffer:TBS, pH 7,3, 0,5% BSA, 0,02% sodium azide.
- Sequence:C-YEHPKDLKLSDGR
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: ELISA: 1:64000; Western Blot: 1:64000
Type: Primary
Antigen: OGT (O-linked N-acetylglucosamine (GlcNAc) transferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human