Anti-Nociceptin Receptor Rabbit Polyclonal Antibody
USBIN2980-02
- Antibody type:Primary
- Antigen name:Nociceptin Receptor
- Antigen symbol:NOP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Nociceptin Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human