Anti-Neuron Specific Enolase Mouse Monoclonal Antibody [clone: 9A115]
Catalog # USBIN2175-02P
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Neuron Specific Enolase
- Antigen symbol:NSE
- Clonality:Monoclonal
- Clone:9A115
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:0,01M PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: Neuron Specific Enolase
Clonality: monoclonal
Clone: 9A115
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human