Anti-Neuron Cell Surface Antigen A2b5 Mouse Monoclonal Antibody (Biotin) [clone: 9H161]
USBIN2172A
- Antibody type:Primary
- Antigen name:Neuron Cell Surface Antigen A2b5
- Clonality:Monoclonal
- Clone:9H161
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgM
- Reactivity:Chicken,Human,Rat,Mouse
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:PBS, containing 50ug BSA.
- Storage temperature:Cool
- Concentration:~500 ug/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Flow Cytometry: 0.1mg/ml
Type: Primary
Antigen: Neuron Cell Surface Antigen A2b5
Clonality: monoclonal
Clone: 9H161
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Chicken, Human, Mouse, Rat