Anti-MHC Class 1, H2, Kd, alloantigen Mouse Monoclonal Antibody [clone: SF1-1.1]
Catalog # USBIM3885-72E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:MHC Class 1, H2, Kd, alloantigen
- Clonality:Monoclonal
- Clone:SF1-1.1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: MHC Class 1, H2, Kd, alloantigen
Clonality: monoclonal
Clone: SF1-1.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse