Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Lysosome Associated Membrane Protein 2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 5K54]
Anti-Lysosome Associated Membrane Protein 2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 5K54]
  USBIL9190-13B
Anti-Lysosome Associated Membrane Protein 2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 5K54]
  USBIL9190-13B

 

  • Antibody type:
    Primary
  • Antigen name:
    Lysosome Associated Membrane Protein 2
  • Antigen symbol:
    LAMP2
  • Clonality:
    Monoclonal
  • Clone:
    5K54
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,1% sodium azide and a stabilizing agent.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Do not freeze!

Type: Primary
Antigen: Lysosome Associated Membrane Protein 2
Clonality: monoclonal
Clone: 5K54
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse