Anti-Lysosome-associated Membrane Protein-1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9A179]
Catalog # USBIL9190-10B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Lysosome Associated Membrane Protein 1
- Antigen symbol:LAMP1
- Clonality:Monoclonal
- Clone:9A179
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide, 0,2% BSA
- Storage temperature:Cool
- Concentration:100 tests/1ml
- Shipping temperature:Ice
- Purification:purified by size exclusion chromatography
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Do not freeze!
Type: Primary
Antigen: Lysosome-associated Membrane Protein-1
Clonality: monoclonal
Clone: 9A179
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human