Anti-Lamin A, C Mouse Monoclonal Antibody [clone: 5F293]
USBIL1220-11B
- Antibody type:Primary
- Antigen name:Lamin A/C
- Clonality:Monoclonal
- Clone:5F293
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgM
- Reactivity:Human,Bovine,Xenopus laevis,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Format:Cell Culture supernatant
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:5 ml
- Pk:5 mL
Type: Primary
Antigen: Lamin A, C
Clonality: monoclonal
Clone: 5F293
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Bovine, Human, Mouse, Rat, Xenopus laevis