Anti-IL10 Rat Monoclonal Antibody [clone: 10B2362]
Catalog # USBII8432-02K
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 10
- Antigen symbol:IL10
- Clonality:Monoclonal
- Clone:10B2362
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:borate buffered saline, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: ; Western Blot: 1:100-1:250; Immunochemistry: 1:100
Type: Primary
Antigen: IL10 (interleukin 10)
Clonality: monoclonal
Clone: 10B2362
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human