Anti-Interferon beta Mouse Monoclonal Antibody [clone: 10D295]
Catalog # USBII7662-10J
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interferon beta
- Antigen symbol:IFNB
- Clonality:Monoclonal
- Clone:10D295
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: Interferon beta
Clonality: monoclonal
Clone: 10D295
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human