Anti-HLA Class 1 Antigen ABC Mouse Monoclonal Antibody [clone: W6/32]
USBIH6098-39R
- Antibody type:Primary
- Antigen name:HLA Class 1 Antigen ABC
- Clonality:Monoclonal
- Clone:W6/32
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Monkey,Human,Bovine,Cat
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: HLA Class 1 Antigen ABC
Clonality: monoclonal
Clone: W6/32
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Bovine, Cat, Human, Monkey