Anti-Histone H1 Mouse monoclonal antibody [clone: 10D158]
Catalog # USBIH5110-01E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Histone H1
- Antigen symbol:H1
- Clonality:Monoclonal
- Clone:10D158
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:100
Type: Primary
Antigen: Histone H1
Clonality: monoclonal
Clone: 10D158
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human