Anti-Glycoprotein 340 Mouse monoclonal antibody [clone: 6F121]
Catalog # USBIG8179-10
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Glycoprotein 340
- Antigen symbol:GP340
- Clonality:Monoclonal
- Clone:6F121
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4,15mM sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1: 200-1:10000
Type: Primary
Antigen: Glycoprotein 340
Clonality: monoclonal
Clone: 6F121
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human