Anti-GASP-2 Mouse monoclonal antibody [clone: 7H70]
Catalog # USBIG2016-76
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:GASP-2
- Clonality:Monoclonal
- Clone:7H70
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Amino acid number:20 to 548
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 25 ?g/ml; Immunoprecipitation: 25 ?g/ml; ELISA: 0.5-1 ?g/ml
Type: Primary
Antigen: GASP-2
Clonality: monoclonal
Clone: 7H70
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human