Anti-FXYD Domain-containing Ion Transport Regulator 1 Rabbit Polyclonal Antibody (Biotin)
USBIF9400-04M-BIOT
- Antibody type:Primary
- Antigen name:FXYD domain-containing ion transport regulator 1
- Antigen symbol:FXYD1
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: FXYD Domain-containing Ion Transport Regulator 1
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse