Anti-Eukaryotic Translation Initiation Factor 4G1 Mouse monoclonal antibody [clone: 10C174]
Catalog # USBIE8949-57F
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Eukaryotic Translation Initiation Factor 4G1
- Clonality:Monoclonal
- Clone:10C174
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:DVAVLKARAKLLQKYLCDEQKELQALYALQALVVTLEQPPNLLRMFFDALYDEDVVKEDAFYSWESSKDPAEQQGKGVALKSVTAFFKWLREAEEESDHN
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:500-1:1000; ELISA: 1 ?g/ml-3ng/ml
Type: Primary
Antigen: Eukaryotic Translation Initiation Factor 4G1
Clonality: monoclonal
Clone: 10C174
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human