Order Entry
United Kingdom
ContactUsLinkComponent
Anti-EPB41L2 Goat polyclonal antibody
Anti-EPB41L2 Goat polyclonal antibody
  USBIE3354-21E
Anti-EPB41L2 Goat polyclonal antibody
  USBIE3354-21E

 

  • Antibody type:
    Primary
  • Antigen name:
    Erythrocyte membrane protein band 4.1-like 2
  • Antigen symbol:
    EPB41L2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
  • Sequence:
    C-DYDPEEHGSID
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: ELISA: 1:16000; Immunochemistry: 2-5 ?g/ml

Type: Primary
Antigen: EPB41L2 (erythrocyte membrane protein band 4.1-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human