Anti-EDIL3 Rabbit Polyclonal Antibody (Biotin)
USBIE2204-30-BIOT
- Antibody type:Primary
- Antigen name:EGF-like repeats and discoidin I-like domains 3
- Antigen symbol:EDIL3
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: EDIL3 (EGF-like repeats and discoidin I-like domains 3)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human