Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Dectin 2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9L821]
Anti-Dectin 2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9L821]
Catalog # USBID1876-49A
Supplier:  US Biological
Anti-Dectin 2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9L821]
Catalog # USBID1876-49A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    dectin 2
  • Clonality:
    Monoclonal
  • Clone:
    9L821
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    liquid
  • Amino acid number:
    46 to 209
  • Storage buffer:
    saline, 0,1% sodium azide, 0,5% BSA.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Do not freeze!

Type: Primary
Antigen: Dectin 2 (dectin 2)
Clonality: monoclonal
Clone: 9L821
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human