Anti-TDGF1 Rabbit Polyclonal Antibody (Biotin)
USBIC7922-11
- Antibody type:Primary
- Antigen name:Teratocarcinoma-derived growth factor 1
- Antigen symbol:TDGF1
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 100ug/ml BSA, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by peptide affinity chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 10?l/Mini blot
Type: Primary
Antigen: TDGF1 (teratocarcinoma-derived growth factor 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse