Anti-COMT Goat Polyclonal Antibody
USBIC7851-70J
- Antibody type:Primary
- Antigen name:catechol-O-methyltransferase
- Antigen symbol:COMT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Form:liquid
- Storage buffer:Tris-buffered saline, pH 7,3, 0,5% BSA, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: ELISA: 1:64000; Western Blot: 0. 03-0.1 ?g/ml; Immunochemistry: 3. 75 ?g/ml
Type: Primary
Antigen: COMT (catechol-O-methyltransferase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human