Anti-CDK6 Mouse monoclonal antibody [clone: 8H4]
Catalog # USBIC2576-01M
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase 6
- Antigen symbol:CDK6
- Clonality:Monoclonal
- Clone:8H4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:3 to 100
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: CDK6 (cyclin-dependent kinase 6)
Clonality: Monoclonal
Clone: 8H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human