Order Entry
United Kingdom
ContactUsLinkComponent
Anti-SELP Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5F52]
Anti-SELP Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5F52]
  USBIC2415-55A
Anti-SELP Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5F52]
  USBIC2415-55A

 

  • Antibody type:
    Primary
  • Antigen name:
    Selectin P
  • Antigen symbol:
    SELP
  • Clonality:
    Monoclonal
  • Clone:
    5F52
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 1% BSA, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    50 Tests
  • Pk:
    50 Tests

 

 

Type: Primary
Antigen: SELP (selectin P (granule membrane protein 140kDa, antigen CD62))
Clonality: Monoclonal
Clone: 5F52
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human