Anti-CDH18 Mouse Monoclonal Antibody [clone: 6F7]
Catalog # USBIC0108-58
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:cadherin 18, type 2
- Antigen symbol:CDH18
- Clonality:Monoclonal
- Clone:6F7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:467 to 577
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: CDH18 (cadherin 18, type 2)
Clonality: Monoclonal
Clone: 6F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human