Anti-Pro-Brain-Derived Neurotrophic Factor Mouse Monoclonal Antibody [clone: 11D34]
Catalog # USBIB2700-10
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Pro-Brain-Derived Neurotrophic Factor
- Clonality:Monoclonal
- Clone:11D34
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:PBS5% trehalose.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0.5-1 ?g/ml; Immunochemistry: 25 ?g/ml
Type: Primary
Antigen: Pro-Brain-Derived Neurotrophic Factor
Clonality: monoclonal
Clone: 11D34
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human