Anti-BMI1 Mouse Monoclonal Antibody [clone: DC9]
USBIB2185-02E
- Antibody type:Primary
- Antigen name:BMI1 polycomb ring finger oncogene
- Antigen symbol:BMI1
- Clonality:Monoclonal
- Clone:DC9
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Format:ascites
- Form:liquid
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 100ug/ml BSA, <0,02% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: BMI1 (BMI1 polycomb ring finger oncogene)
Clonality: monoclonal
Clone: DC9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Monkey, Mouse, Rat