Anti-Aprotinin Mouse Monoclonal Antibody (Biotin) [clone: 9A509]
USBIA2300-01B-50
- Antibody type:Primary
- Antigen name:Aprotinin
- Clonality:Monoclonal
- Clone:9A509
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Bovine
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBSpH 7,4, 15mM sodium azide
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µl
- Pk:50 µl
Recommended dilution: ELISA: 1:20000
Type: Primary
Antigen: Aprotinin
Clonality: monoclonal
Clone: 9A509
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine