Anti-APOF Mouse monoclonal antibody unconjugated [clone: 1D5]
Catalog # USBIA2299-76V1
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Apolipoprotein F
- Antigen symbol:APOF
- Clonality:Monoclonal
- Clone:1D5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: APOF (apolipoprotein F)
Clonality: monoclonal
Clone: 1D5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human