Anti-Adenovirus hexon Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 9L8]
Catalog # USBIA0880-04B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Adenovirus Hexon
- Clonality:Monoclonal
- Clone:9L8
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:
- Environmentally Preferable:
- Form:liquid
- Storage buffer:0,01M PBS, pH 7,2, 10mg/ml BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Adenovirus hexon
Clonality: monoclonal
Clone: 9L8
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: