Anti-CCR5 Mouse Monoclonal Antibody [clone: 14A2]
Catalog # USBI167722
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) receptor 5(gene/pseudogene)
- Antigen symbol:CCR5
- Clonality:Monoclonal
- Clone:14A2
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:200-1:400
Type: Primary
Antigen: CCR5 (chemokine (C-C motif) receptor 5 (gene/pseudogene))
Clonality: monoclonal
Clone: 14A2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: