Anti-ASIC1 Mouse Monoclonal Antibody [clone: S271-44]
Catalog # USBI167577
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Acid sensing (proton gated) ion channel 1
- Antigen symbol:ASCI1
- Clonality:Monoclonal
- Clone:S271-44
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Antigen synonyms:ACCN2
- Storage buffer:PBSpH 7,4, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ASIC1 (acid-sensing (proton-gated) ion channel 1)
Clonality: monoclonal
Clone: S271-44
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat