Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-MAT2B Goat Polyclonal Antibody
Catalog # USBI167447
Supplier:  US Biological
Anti-MAT2B Goat Polyclonal Antibody
Catalog # USBI167447
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Methionine adenosyltransferase 2 beta
  • Antigen symbol:
    MAT2B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    Tris saline, pH 7,3, 0,5% BSA, 0,02% sodium azide.
  • Sequence:
    REKELSIHFVPGSC
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by ammonium sulfate precipitation
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: ELISA: 1:128000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: MAT2B (methionine adenosyltransferase II, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse