Order Entry
United Kingdom
ContactUsLinkComponent
Anti-DCLK1 Rabbit Polyclonal Antibody
Anti-DCLK1 Rabbit Polyclonal Antibody
  USBI146609
Anti-DCLK1 Rabbit Polyclonal Antibody
  USBI146609

 

  • Antibody type:
    Primary
  • Antigen name:
    doublecortin-like kinase 1
  • Antigen symbol:
    DCLK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: DCLK1 (doublecortin-like kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat