Anti-Protocadherin-15 Mouse Monoclonal Antibody [clone: 740523]
Catalog # USBI145582
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Protocadherin-15
- Clonality:Monoclonal
- Clone:740523
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:PBS,Trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 825 ?g/ml
Type: Primary
Antigen: Protocadherin-15
Clonality: monoclonal
Clone: 740523
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human