Anti-Glutathione S Transferase, pi 1 Mouse Monoclonal Antibody [clone: 800027]
Catalog # USBI145479
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Glutathione S-transferase pi 1
- Antigen symbol:GSTP1
- Clonality:Monoclonal
- Clone:800027
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:PBS,Trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.25 ?g/ml; Immunochemistry: 825 ?g/ml
Type: Primary
Antigen: Glutathione S Transferase, pi 1
Clonality: monoclonal
Clone: 800027
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse