Anti-Carbohydrate Sulfotransferase 3 Mouse Monoclonal Antibody [clone: 799011]
USBI145405
- Antibody type:Primary
- Antigen name:Carbohydrate Sulfotransferase 3
- Antigen symbol:CPS3
- Clonality:Monoclonal
- Clone:799011
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:PBS,Trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Immunochemistry: 825 ?g/ml
Type: Primary
Antigen: Carbohydrate Sulfotransferase 3
Clonality: monoclonal
Clone: 799011
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human