Anti-CDH5 Rabbit Polyclonal Antibody
USBI144347
- Antibody type:Primary
- Antigen name:cadherin 5, type 2 (vascular endothelium)
- Antigen symbol:CDH5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:5mgBSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mgthimerosal, 0,05mg NaN3.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: CDH5 (cadherin 5, type 2 (vascular endothelium))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat