Anti-YY1AP1 Mouse Monoclonal Antibody [clone: 13G80]
Catalog # USBI137510
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:YY1 associated protein 1
- Antigen symbol:YY1AP1
- Clonality:Monoclonal
- Clone:13G80
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Hamster,Monkey,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM NaCl, 100ug/ml BSA, 50% glycerol and <0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:400; Immunoprecipitation: 1:200
Type: Primary
Antigen: YY1AP1 (YY1 associated protein 1)
Clonality: monoclonal
Clone: 13G80
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Hamster, Human, Monkey, Mouse, Rat