Anti-CA15-3 Cancer Antigen Mouse Monoclonal Antibody [clone: 13F7]
USBI136549
- Antibody type:Primary
- Antigen name:CA15-3 Cancer Antigen
- Antigen symbol:CA15-3
- Clonality:Monoclonal
- Clone:13F7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human,Mouse
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Recommended dilution: ELISA:; Western Blot: 1:10000
Type: Primary
Antigen: CA15-3 Cancer Antigen
Clonality: monoclonal
Clone: 13F7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse