Anti-VAPB Mouse Monoclonal Antibody [clone: 1A2]
USBI135208
- Antibody type:Primary
- Antigen name:VAMP (vesicle-associated membrane Protein)-associated protein B
- Antigen symbol:VAPB
- Clonality:Monoclonal
- Clone:1A2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:124 to 224
- Storage buffer:PBS, pH 7,2.
- Sequence:ELPAENDKPHDVEINKIISTTASKTETPIVSKSLSSSLDDTEVKKVMEECKRLQGEVQRLREENKQFKEEDGLRMRKTVQSNSPISALAPTGKEEGLSTR
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: VAPB (VAMP (vesicle-associated membrane protein)-associated protein B and C)
Clonality: monoclonal
Clone: 1A2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human