Anti-TCEB3 Mouse Monoclonal Antibody [clone: 1F3]
Catalog # USBI134285
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Transcription Elongation Factor B (SIII), Polypeptide 3
- Antigen symbol:TCEB3
- Clonality:Monoclonal
- Clone:1F3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:81 to 191
- Storage buffer:PBS, pH 7,2.
- Sequence:NAEPDEQDFEKSNSRKRPRDALQKEEEMEGDYQETWKATGSRSYSPDHRQKKHRKLSELERPHKVSHGHERRDERKRCHRMSPTYSSDPESSDYGHVQSPPSCTSPHQMY
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: TCEB3 (transcription elongation factor B (SIII), polypeptide 3 (110kDa, elongin A))
Clonality: monoclonal
Clone: 1F3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human