Anti-SLC25A13 Rabbit Polyclonal Antibody
Catalog # USBI133430
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 25 (aspartate/glutamate carrier), member 13
- Antigen symbol:Solute carrier family 25, member 13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:1 to 675
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SLC25A13 (solute carrier family 25 (aspartate/glutamate carrier), member 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse